Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002365.4 | NP_002356.2 | 5 | 957 | ||
XM_011545513.2 | XP_011543815.1 | 4 | 564 | |||
XM_011545514.2 | XP_011543816.1 | 3 | 531 | |||
GenBank mRNA | AK057441.1 | - | 5 | 957 | ||
BC069784.1 | - | 1 | 279 | 7262601 | ||
BC074756.2 | - | 1 | 248 | 30915215 | ||
BC126320.1 | - | 1 | 251 | 8992036 | ||
BC126322.1 | - | 1 | 251 | 8992038 |