Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002373.5 | NP_002364.5 | 4 | 757 | ||
XM_017022189.1 | XP_016877678.1 | 5 | 1047 | |||
GenBank mRNA | AF200415.2 | - | 2 | 489 | ||
AK291052.1 | - | 4 | 729 | |||
BC055312.1 | - | 4 | 726 | |||
BC094702.1 | - | 4 | 716 | |||
U38292.1 | - | 1 | 290 | |||
U80458.1 | - | 1 | 59 |