Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002386.3 | NP_002377.4 | 1 | 1918 | ||
GenBank mRNA | AK024232.1 | - | 1 | 1920 | ||
AK054666.1 | - | 2 | 888 | |||
AK225902.1 | - | 1 | 935 | |||
AK311630.1 | - | 2752 | ||||
BC007856.2 | - | 1 | 917 | 4298428 | ||
BC080622.1 | - | 1 | 744 | 6267157 | ||
X67594.1 | - | 1 | 706 |
Set Membership: |
![]() |