Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085377.1 | NP_001078846.1 | 19 | 4247 | ||
NM_002387.2 | NP_002378.1 | 17 | 3791 | |||
XM_005271991.2 | XP_005272048.1 | 18 | 3891 | |||
XM_017009473.1 | XP_016864962.1 | 20 | 3912 | |||
XM_017009474.1 | XP_016864963.1 | 18 | 6577 | |||
GenBank mRNA | BX537952.1 | - | 18 | 3262 | ||
BX647710.1 | - | 20 | 4183 | |||
M62397.1 | - | 18 | 3481 |