Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002388.4 | NP_002379.3 | 2 | 299 | ||
GenBank mRNA | AK222586.1 | - | 2 | 137 | ||
AK301704.1 | - | 2 | 238 | |||
AK312928.1 | - | 2 | 108 | |||
AY032603.1 | - | 2 | 145 | |||
BC001626.1 | - | 2 | 144 | 3542098 | ||
BC003509.1 | - | 2 | 145 | 3610786 | ||
D38073.1 | - | 2 | 169 | |||
KU178170.1 | - | 2 | 92 | |||
KU178171.1 | - | 2 | 92 | |||
X62153.2 | - | 2 | 136 |