Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270472.1 | NP_001257401.1 | 16 | 3044 | ||
NM_002388.4 | NP_002379.3 | 17 | 3157 | |||
GenBank mRNA | AK222586.1 | - | 17 | 2995 | ||
AY032603.1 | - | 17 | 3004 | |||
BC001626.1 | - | 17 | 3002 | 3542098 | ||
BC003509.1 | - | 17 | 3003 | 3610786 | ||
D38073.1 | - | 17 | 3027 | |||
X62153.2 | - | 17 | 2994 |