Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145938.1 | NP_001139410.1 | 4, 5 | 610 | ||
NM_002421.3 | NP_002412.1 | 4, 5 | 762 | |||
GenBank mRNA | AK024818.1 | - | 4, 5 | 685 | ||
AK098450.1 | - | 5, 6 | 617 | |||
AK130687.1 | - | 4, 5 | 685 | |||
AK223034.1 | - | 3, 4 | 388 | |||
AK223035.1 | - | 4, 5 | 643 | |||
AK223036.1 | - | 2, 3 | 226 | |||
AK223056.1 | - | 3, 4 | 428 | |||
AK223281.1 | - | 4, 5 | 685 | |||
AK225495.1 | - | 3, 4 | 453 | |||
AK297723.1 | - | 4, 5 | 533 | |||
AK301340.1 | - | 4, 5 | 475 | |||
BC013875.2 | - | 4, 5 | 661 | 3834572 | ||
BT006874.1 | - | 4, 5 | 619 | |||
BT020147.1 | - | 4, 5 | 619 | |||
M13509.1 | - | 4, 5 | 687 | |||
X54925.1 | - | 4, 5 | 687 |