Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002442.3 | NP_002433.1 | 4 | 346 | ||
XM_006719403.3 | XP_006719466.1 | 4 | 340 | |||
XM_011538361.2 | XP_011536663.1 | 4 | 340 | |||
XM_011538362.2 | XP_011536664.1 | 4 | 340 | |||
XM_011538363.2 | XP_011536665.1 | 4 | 340 | |||
GenBank mRNA | AB012851.1 | - | 4 | 321 | ||
AB072590.1 | - | 1 | 68 | |||
AB072591.1 | - | 1 | 68 | |||
AB072592.1 | - | 1 | 68 | |||
AB073212.1 | - | 1 | 61 | |||
BX393884.2 | - | 4 | 346 |