Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002444.2 | NP_002435.1 | 6, 7 | 893 | ||
XM_005262269.2 | XP_005262326.1 | 6, 7 | 914 | |||
XM_011530959.1 | XP_011529261.1 | 6, 7 | 809 | |||
XM_017029545.1 | XP_016885034.1 | 8, 9 | 1037 | |||
XM_017029546.1 | XP_016885035.1 | 7, 8 | 933 | |||
GenBank mRNA | BC001112.2 | - | 6, 7 | 865 | ||
BC011827.2 | - | 6, 7 | 865 | |||
BC017293.2 | - | 6, 7 | 846 | 4908580 | ||
EU794636.1 | - | 6, 7 | 846 | |||
M69066.1 | - | 6, 7 | 795 |