Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MST1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244937.2 | NP_001231866.1 | 1 | 805 | ||
NM_001318913.1 | NP_001305842.1 | 1 | 805 | |||
NM_002447.3 | NP_002438.2 | 1 | 805 | |||
NR_134919.1 | 1 | 805 | ||||
XM_005265170.4 | XP_005265227.2 | 1 | 800 | |||
XM_011533739.2 | XP_011532041.1 | 1 | 800 | |||
XM_011533740.2 | XP_011532042.1 | 1 | 800 | |||
XM_011533741.2 | XP_011532043.1 | 1 | 800 | |||
XM_011533742.2 | XP_011532044.1 | 1 | 800 | |||
XM_011533743.2 | XP_011532045.1 | 1 | 800 | |||
XM_011533744.2 | XP_011532046.1 | 1 | 800 | |||
GenBank mRNA | EU826582.1 | - | 1 | 777 | ||
EU826583.1 | - | 1 | 777 | |||
EU826584.1 | - | 1 | 777 | |||
EU826585.1 | - | 1 | 777 | |||
HQ908652.1 | - | 1 | 777 | |||
JN983806.1 | - | 1 | 805 | |||
X70040.1 | - | 1 | 805 |