Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 8 | 1318 | ||
NM_001321001.1 | NP_001307930.1 | 7 | 853 | |||
NM_001321002.1 | NP_001307931.1 | 7 | 825 | |||
NM_001321003.1 | NP_001307932.1 | 8 | 1035 | |||
NM_001321004.1 | NP_001307933.1 | 6 | 1087 | |||
NM_002453.2 | NP_002444.2 | 7 | 897 | |||
XM_005264335.3 | XP_005264392.1 | 5 | 696 | |||
XM_011532871.2 | XP_011531173.1 | 6 | 730 | |||
XM_017004162.1 | XP_016859651.1 | 8 | 1026 | |||
XM_017004163.1 | XP_016859652.1 | 7 | 1102 | |||
XM_017004164.1 | XP_016859653.1 | 7 | 891 | |||
XM_017004167.1 | XP_016859656.1 | 3 | 342 | |||
GenBank mRNA | AF494407.1 | - | 7 | 877 | ||
AK225184.1 | - | 7 | 866 | |||
AK307993.1 | - | 7 | 877 | |||
BC039848.1 | - | 7 | 855 | |||
BC064977.1 | - | 7 | 837 | |||
L34600.1 | - | 7 | 851 |