Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 11 | 1814 | ||
NM_001321001.1 | NP_001307930.1 | 10 | 1349 | |||
NM_001321002.1 | NP_001307931.1 | 10 | 1321 | |||
NM_001321003.1 | NP_001307932.1 | 11 | 1531 | |||
NM_001321004.1 | NP_001307933.1 | 9 | 1583 | |||
NM_001321005.1 | NP_001307934.1 | 9 | 1232 | |||
NM_002453.2 | NP_002444.2 | 10 | 1393 | |||
XM_005264335.3 | XP_005264392.1 | 8 | 1192 | |||
XM_011532871.2 | XP_011531173.1 | 9 | 1226 | |||
XM_017004162.1 | XP_016859651.1 | 11 | 1522 | |||
XM_017004163.1 | XP_016859652.1 | 10 | 1598 | |||
XM_017004164.1 | XP_016859653.1 | 9 | 1210 | |||
XM_017004165.1 | XP_016859654.1 | 10 | 1425 | |||
XM_017004166.1 | XP_016859655.1 | 10 | 1617 | |||
XM_017004167.1 | XP_016859656.1 | 6 | 1694 | |||
GenBank mRNA | AF494407.1 | - | 10 | 1373 | ||
AK225184.1 | - | 10 | 1362 | |||
AK307993.1 | - | 10 | 1373 | |||
BC039848.1 | - | 10 | 1351 | |||
BC064977.1 | - | 10 | 1333 | |||
L34600.1 | - | 10 | 1347 |