Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 15 | 1837 | ||
NM_002498.2 | NP_002489.1 | 16 | 1888 | |||
NM_152720.2 | NP_689933.1 | 16 | 1907 | |||
NR_027415.2 | 16 | 1887 | ||||
GenBank mRNA | AB072828.1 | - | 14 | 1506 | ||
AK131359.1 | - | 14 | 2755 | |||
AK290259.1 | - | 16 | 1620 | |||
AL833951.1 | - | 15 | 5270 | |||
BC019916.1 | - | 16 | 1615 | 5000918 | ||
Z29067.1 | - | 14 | 1371 |