Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002515.2 | NP_002506.2 | 3 | 640 | ||
NM_006489.2 | NP_006480.2 | 3 | 640 | |||
NM_006491.2 | NP_006482.1 | 3 | 640 | |||
XM_011536800.2 | XP_011535102.1 | 3 | 208 | |||
XM_017021342.1 | XP_016876831.1 | 3 | 696 | |||
XM_017021343.1 | XP_016876832.1 | 3 | 696 | |||
XM_017021344.1 | XP_016876833.1 | 4 | 507 | |||
XM_017021345.1 | XP_016876834.1 | 3 | 854 | |||
XM_017021346.1 | XP_016876835.1 | 4 | 956 | |||
XM_017021347.1 | XP_016876836.1 | 4 | 837 | |||
XM_017021348.1 | XP_016876837.1 | 3 | 772 | |||
XM_017021349.1 | XP_016876838.1 | 3 | 855 | |||
GenBank mRNA | AK226114.1 | - | 4 | 459 | ||
AK289639.1 | - | 3 | 326 | |||
AK291022.1 | - | 3 | 463 | |||
AK291468.1 | - | 3 | 341 | |||
AK301526.1 | - | 4 | 424 | |||
BC075038.2 | - | 3 | 345 | 30915356 | ||
BC075039.2 | - | 3 | 346 | 30915572 | ||
CN409914.1 | - | 3 | 643 | |||
U04840.1 | - | 3 | 391 |