Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321307.1 | NP_001308236.1 | 11 | 1041 | ||
NM_002519.2 | NP_002510.2 | 11 | 1041 | |||
XM_011542854.2 | XP_011541156.1 | 11 | 1041 | |||
XM_017017802.1 | XP_016873291.1 | 11 | 1779 | |||
XM_017017803.1 | XP_016873292.1 | 11 | 1222 | |||
GenBank mRNA | AK290020.1 | - | 11 | 1041 | ||
AK291687.1 | - | 11 | 965 | |||
BC040356.1 | - | 11 | 1032 | |||
BC050561.2 | - | 11 | 1026 | |||
BC136270.1 | - | 11 | 962 | 9020257 | ||
D83243.1 | - | 11 | 1005 | |||
U58852.1 | - | 11 | 939 | |||
X97186.1 | - | 11 | 973 |