Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002531.2 | NP_002522.2 | 4 | 3914 | ||
GenBank mRNA | BC040711.1 | - | 1 | 1234 | ||
X70070.1 | - | 4 | 3914 |
Set Membership: |
G-protein coupled receptor |