Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320653.1 | NP_001307582.1 | 1 | 196 | ||
NM_002532.5 | NP_002523.2 | 1 | 196 | |||
XM_011523893.2 | XP_011522195.1 | 1 | 119 | |||
XM_017024694.1 | XP_016880183.1 | 1 | 119 | |||
GenBank mRNA | AK225247.1 | - | 1 | 119 | ||
AK299000.1 | - | 1 | 119 | |||
BC000335.2 | - | 1 | 109 | 2822595 | ||
BI552947.1 | - | 1 | 138 | |||
BX956129.1 | - | 1 | 196 | |||
Y08612.2 | - | 1 | 155 |