Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320653.1 | NP_001307582.1 | 9 | 1402 | ||
NM_002532.5 | NP_002523.2 | 9 | 1402 | |||
XM_011523893.2 | XP_011522195.1 | 10 | 1452 | |||
XM_017024694.1 | XP_016880183.1 | 10 | 1452 | |||
GenBank mRNA | AK225247.1 | - | 9 | 1322 | ||
AK298158.1 | - | 10 | 1179 | |||
AK299000.1 | - | 9 | 1325 | |||
AK308142.1 | - | 9 | 1214 | |||
BC000335.2 | - | 9 | 1315 | 2822595 | ||
Y08612.2 | - | 9 | 1361 |