Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143666
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
12, 13
1670
NM_001242353.1
NP_001229282.1
12, 13
1756
NM_001242354.1
NP_001229283.1
11, 12
1442
NM_002540.4
NP_002531.3
14, 15
1958
NM_153432.1
NP_702910.1
11, 12
1516
NM_153433.1
NP_702911.1
12, 13
1683
NM_153435.1
NP_702913.1
12, 13
1576
NM_153436.1
NP_702914.1
11, 12
1340
NM_153437.2
NP_702915.1
12, 13
1606
NM_153439.1
NP_702917.1
11, 12
1459
NM_153440.1
NP_702918.1
11, 12
1386
XM_005252010.2
XP_005252067.1
11, 12
1468
XM_005252011.2
XP_005252068.1
11, 12
1408
XM_005252012.3
XP_005252069.1
12, 13
1658
XM_005252013.3
XP_005252070.1
12, 13
1605
XM_005252014.3
XP_005252071.1
12, 13
1654
XM_005252017.4
XP_005252074.1
12, 13
1559
XM_005252018.2
XP_005252075.1
13, 14
1764
XM_005252019.3
XP_005252076.1
13, 14
1783
XM_005252022.2
XP_005252079.1
12, 13
1582
XM_005252023.2
XP_005252080.1
12, 13
1553
XM_005252024.3
XP_005252081.1
12, 13
1572
XM_005252026.4
XP_005252083.1
12, 13
1501
XM_005252029.2
XP_005252086.1
12, 13
1582
XM_006717126.3
XP_006717189.1
12, 13
1658
XM_011518722.2
XP_011517024.1
12, 13
1582
XM_011518723.2
XP_011517025.1
12, 13
1525
XM_011518724.2
XP_011517026.1
13, 14
1746
XM_011518725.2
XP_011517027.1
13, 14
1750
XM_011518726.2
XP_011517028.1
13, 14
1768
XM_011518728.2
XP_011517030.1
12, 13
1519
XM_011518729.2
XP_011517031.1
12, 13
1455
XM_011518730.2
XP_011517032.1
12, 13
1582
XM_011518731.2
XP_011517033.1
11, 12
1411
XM_011518732.2
XP_011517034.1
12, 13
1368
XM_011518733.2
XP_011517035.1
11, 12
1354
XM_011518744.1
XP_011517046.1
11, 12
1408
XM_017014764.1
XP_016870253.1
13, 14
1739
XM_017014765.1
XP_016870254.1
13, 14
1758
XM_017014766.1
XP_016870255.1
11, 12
1468
XM_017014767.1
XP_016870256.1
13, 14
1750
XM_017014768.1
XP_016870257.1
12, 13
1542
XM_017014769.1
XP_016870258.1
12, 13
1571
XM_017014770.1
XP_016870259.1
13, 14
1726
XM_017014771.1
XP_016870260.1
13, 14
1707
XM_017014772.1
XP_016870261.1
13, 14
1783
XM_017014773.1
XP_016870262.1
13, 14
1759
XM_017014774.1
XP_016870263.1
12, 13
1469
XM_017014775.1
XP_016870264.1
13, 14
1726
XM_017014776.1
XP_016870265.1
13, 14
1707
XM_017014777.1
XP_016870266.1
12, 13
1509
XM_017014778.1
XP_016870267.1
12, 13
1612
XM_017014779.1
XP_016870268.1
11, 12
1353
XM_017014780.1
XP_016870269.1
12, 13
1536
XM_017014781.1
XP_016870270.1
11, 12
1291
GenBank mRNA
AK093367.1
-
11, 12
1466
AK299303.1
-
12, 13
1518
AK300793.1
-
11, 12
1291
AK301754.1
-
11, 12
1383
AK301842.1
-
11, 12
1308
AK301855.1
-
11, 12
1362
AK302684.1
-
12, 13
1610
AK308278.1
-
10, 11
1222
AY319414.1
-
11, 12
1492
AY366499.1
-
12, 13
1453
BC010629.1
-
14, 15
1756
3874501
BC091500.1
-
12, 13
1570
30378678
DQ444713.1
-
10, 11
1283
DQ444714.1
-
10, 11
1226
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline