Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143668
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
16
2038
NM_001242353.1
NP_001229282.1
16
2124
NM_001242354.1
NP_001229283.1
15
1810
NM_002540.4
NP_002531.3
18
2326
NM_153432.1
NP_702910.1
15
1884
NM_153433.1
NP_702911.1
16
2051
NM_153435.1
NP_702913.1
16
1944
NM_153436.1
NP_702914.1
15
1708
NM_153437.2
NP_702915.1
16
1974
NM_153439.1
NP_702917.1
15
1827
NM_153440.1
NP_702918.1
15
1754
XM_005252010.2
XP_005252067.1
15
1836
XM_005252011.2
XP_005252068.1
15
1776
XM_005252012.3
XP_005252069.1
16
2026
XM_005252013.3
XP_005252070.1
16
1973
XM_005252014.3
XP_005252071.1
16
2022
XM_005252017.4
XP_005252074.1
16
1927
XM_005252018.2
XP_005252075.1
17
2132
XM_005252019.3
XP_005252076.1
17
2151
XM_005252022.2
XP_005252079.1
16
1950
XM_005252023.2
XP_005252080.1
16
1921
XM_005252024.3
XP_005252081.1
16
1940
XM_005252026.4
XP_005252083.1
16
1869
XM_005252029.2
XP_005252086.1
16
1950
XM_006717126.3
XP_006717189.1
16
2026
XM_011518722.2
XP_011517024.1
16
1950
XM_011518723.2
XP_011517025.1
16
1893
XM_011518724.2
XP_011517026.1
17
2114
XM_011518725.2
XP_011517027.1
17
2118
XM_011518726.2
XP_011517028.1
17
2136
XM_011518728.2
XP_011517030.1
16
1887
XM_011518729.2
XP_011517031.1
16
1823
XM_011518730.2
XP_011517032.1
16
1950
XM_011518731.2
XP_011517033.1
15
1779
XM_011518732.2
XP_011517034.1
16
1736
XM_011518733.2
XP_011517035.1
15
1722
XM_011518744.1
XP_011517046.1
15
1776
XM_017014764.1
XP_016870253.1
17
2107
XM_017014765.1
XP_016870254.1
17
2126
XM_017014766.1
XP_016870255.1
15
1836
XM_017014767.1
XP_016870256.1
17
2118
XM_017014768.1
XP_016870257.1
16
1910
XM_017014769.1
XP_016870258.1
16
1939
XM_017014770.1
XP_016870259.1
17
2094
XM_017014771.1
XP_016870260.1
17
2075
XM_017014772.1
XP_016870261.1
17
2151
XM_017014773.1
XP_016870262.1
17
2127
XM_017014774.1
XP_016870263.1
16
1837
XM_017014775.1
XP_016870264.1
17
2094
XM_017014776.1
XP_016870265.1
17
2075
XM_017014777.1
XP_016870266.1
16
1877
XM_017014778.1
XP_016870267.1
16
1980
XM_017014779.1
XP_016870268.1
15
1721
XM_017014780.1
XP_016870269.1
16
1904
XM_017014781.1
XP_016870270.1
15
1659
GenBank mRNA
AF012549.1
-
16
1914
AF053970.1
-
14
1613
AK093367.1
-
15
1834
AK299303.1
-
16
1886
AK300793.1
-
15
1659
AK301754.1
-
15
1751
AK301842.1
-
15
1676
AK301855.1
-
15
1730
AK302684.1
-
16
1978
AY319414.1
-
15
1860
AY366499.1
-
16
1821
BC010629.1
-
18
2124
3874501
BC091500.1
-
16
1938
30378678
DB511337.1
-
2
39
DQ444713.1
-
14
1651
DQ444714.1
-
14
1594
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0