Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 8 | 2075 | ||
NM_001319103.1 | NP_001306032.1 | 8 | 2380 | |||
NM_001319104.1 | NP_001306033.1 | 7 | 1990 | |||
NM_001319105.1 | NP_001306034.1 | 8 | 1899 | |||
NM_001319106.1 | NP_001306035.1 | 7 | 2350 | |||
NM_002545.4 | NP_002536.1 | 7 | 2353 | |||
XM_006718846.2 | XP_006718909.1 | 8 | 2298 | |||
XM_011542856.2 | XP_011541158.1 | 9 | 2611 | |||
GenBank mRNA | BC117254.1 | - | 7 | 1830 | 40125805 | |
BC126251.1 | - | 7 | 1831 | 8991967 | ||
BC143945.1 | - | 7 | 1828 | 9052463 | ||
BC143946.1 | - | 8 | 1858 | 9052464 | ||
BX537377.1 | - | 8 | 1923 | |||
U79251.1 | - | 1 | 585 |