Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004279.2 | NP_004270.2 | 2 | 180 | ||
XM_005250717.3 | XP_005250774.1 | 2 | 194 | |||
XM_006716181.3 | XP_006716244.1 | 2 | 193 | |||
GenBank mRNA | AF054182.1 | - | 2 | 159 | ||
AK000994.1 | - | 2 | 162 | |||
AK057656.1 | - | 2 | 172 | |||
AK098616.1 | - | 2 | 150 | |||
AK289864.1 | - | 2 | 162 | |||
AK297354.1 | - | 2 | 545 | |||
AK309928.1 | - | 150 | ||||
BC014079.2 | - | 2 | 120 | |||
BP311561.1 | - | 2 | 178 |