Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002557.3 | NP_002548.3 | 11 | 1731 | ||
GenBank mRNA | AB208783.1 | - | 9 | 2113 | ||
AK298757.1 | - | 9 | 1474 | |||
AK301726.1 | - | 7 | 1213 | |||
AK314074.1 | - | 11 | 1689 | |||
AY189737.1 | - | 11 | 1732 | |||
BC126177.1 | - | 11 | 1714 | 8991893 | ||
BC136406.1 | - | 11 | 1714 | 9020393 | ||
U09550.1 | - | 11 | 1687 |