Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002565.3 | NP_002556.1 | 1 | 1014 | ||
GenBank mRNA | BC095503.1 | - | 1 | 973 | 30706597 | |
BC096067.3 | - | 1 | 877 | 40000385 | ||
BC096068.1 | - | 1 | 877 | 40000387 | ||
BC096069.1 | - | 1 | 877 | 40000391 | ||
BC096070.1 | - | 1 | 877 | 40000394 |
Set Membership: |
G-protein coupled receptor |