Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002565.3 | NP_002556.1 | 1 | 1344 | ||
GenBank mRNA | BC095503.1 | - | 1 | 1303 | 30706597 |
Set Membership: |
G-protein coupled receptor |