Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000906.3 | NP_000897.3 | 1, 2 | 1144 | ||
XM_005245218.1 | XP_005245275.1 | 1, 2 | 1215 | |||
XM_017001374.1 | XP_016856863.1 | 1, 2 | 1215 | |||
GenBank mRNA | AK025024.1 | - | 1, 2 | 1144 | ||
BC063304.1 | - | 1, 2 | 1193 | 30331757 | ||
X15357.1 | - | 1, 2 | 766 |