Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143709
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
5
784
NM_001128167.2
NP_001121639.1
4
711
NM_001128168.2
NP_001121640.1
9
1033
NM_001128172.2
NP_001121644.1
4
386
NM_001128173.2
NP_001121645.1
8
970
NM_001324325.1
NP_001311254.1
5
862
NM_001324326.1
NP_001311255.1
5
707
NM_001324327.1
NP_001311256.1
7
819
NM_001324328.1
NP_001311257.1
6
752
NM_001324329.1
NP_001311258.1
8
967
NM_001324330.1
NP_001311259.1
5
859
NM_001324331.1
NP_001311260.1
6
774
NM_001324332.1
NP_001311261.1
7
925
NM_001324333.1
NP_001311262.1
7
900
NM_001324334.1
NP_001311263.1
6
855
NM_002578.4
NP_002569.1
7
922
NR_136747.1
6
774
NR_136748.1
4
471
XM_005262132.2
XP_005262189.1
8
962
XM_005262133.3
XP_005262190.1
7
917
XM_006724654.2
XP_006724717.1
7
809
XM_006724655.2
XP_006724718.1
6
752
XM_011530962.1
XP_011529264.1
8
960
XM_011530967.2
XP_011529269.1
7
900
XM_011530968.2
XP_011529270.1
8
976
XM_011530969.1
XP_011529271.1
6
821
XM_011530971.2
XP_011529273.1
8
962
XM_017029557.1
XP_016885046.1
6
821
XM_017029558.1
XP_016885047.1
6
764
XM_017029559.1
XP_016885048.1
5
776
XM_017029560.1
XP_016885049.1
5
707
XM_017029561.1
XP_016885050.1
7
924
XM_017029562.1
XP_016885051.1
8
962
XM_017029563.1
XP_016885052.1
9
1025
GenBank mRNA
AB102659.1
-
4
341
AF068864.1
-
3
296
AF155651.1
-
7
687
AK128022.1
-
6
1010
AK290504.1
-
4
711
AK315284.1
-
5
628
AM943850.1
-
4
341
AM943851.1
-
4
359
AM943852.1
-
5
404
BC117353.1
-
4
390
BC117355.1
-
4
390
BC143544.1
-
4
390
EF491818.1
-
3
114
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0