Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040134.1 | NP_001035224.1 | 8 | 1337 | ||
NM_002579.2 | NP_002570.2 | 9 | 1469 | |||
XM_005259565.4 | XP_005259622.1 | 9 | 1485 | |||
XM_005259566.4 | XP_005259623.1 | 9 | 1828 | |||
XM_017026850.1 | XP_016882339.1 | 8 | 1376 | |||
GenBank mRNA | AK022919.1 | - | 5 | 1013 | ||
AK301181.1 | - | 6 | 1085 | |||
BC019596.1 | - | 7 | 1168 | |||
BC032449.1 | - | 9 | 1317 | 5179938 | ||
CN344023.1 | - | 1 | 330 | |||
D87460.1 | - | 6 | 1149 | |||
Y14770.1 | - | 9 | 1302 |