Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002579.2 | NP_002570.2 | 8, 9 | 826 | ||
XM_005259565.4 | XP_005259622.1 | 8, 9 | 842 | |||
XM_005259566.4 | XP_005259623.1 | 8, 9 | 1185 | |||
GenBank mRNA | AK301181.1 | - | 5, 6 | 442 | ||
AK315168.1 | - | 8, 9 | 678 | |||
BC032449.1 | - | 8, 9 | 674 | 5179938 | ||
D87460.1 | - | 5, 6 | 506 | |||
Y14770.1 | - | 8, 9 | 659 |