Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004292.2 | NP_004283.2 | 10 | 2647 | ||
XM_011545399.2 | XP_011543701.1 | 10 | 2996 | |||
XM_011545400.2 | XP_011543702.1 | 10 | 2366 | |||
XM_011545401.2 | XP_011543703.1 | 10 | 2521 | |||
XM_017018587.1 | XP_016874076.1 | 10 | 2369 | |||
GenBank mRNA | AB209445.1 | - | 7 | 2550 | ||
AK297757.1 | - | 10 | 2060 | |||
BC014417.1 | - | 10 | 2537 | 4549333 | ||
L36463.1 | - | 10 | 2647 |