Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242339.1 | NP_001229268.1 | 24 | 2636 | ||
NM_001323067.1 | NP_001309996.1 | 22 | 2375 | |||
NM_001323068.1 | NP_001309997.1 | 21 | 2359 | |||
NM_001323069.1 | NP_001309998.1 | 22 | 2507 | |||
NM_001323070.1 | NP_001309999.1 | 16 | 2169 | |||
NM_001323071.1 | NP_001310000.1 | 23 | 2628 | |||
NM_001323072.1 | NP_001310001.1 | 23 | 2735 | |||
NM_001323073.1 | NP_001310002.1 | 18 | 1996 | |||
NM_001323074.1 | NP_001310003.1 | 17 | 2322 | |||
NM_002627.4 | NP_002618.1 | 22 | 2512 | |||
XM_005252465.4 | XP_005252522.1 | 23 | 2710 | |||
XM_005252466.4 | XP_005252523.1 | 22 | 2557 | |||
XM_005252467.2 | XP_005252524.1 | 22 | 2481 | |||
XM_006717449.1 | XP_006717512.1 | 23 | 2634 | |||
GenBank mRNA | AK092597.1 | - | 24 | 2636 | ||
AK097692.1 | - | 6 | 1565 | |||
AK128373.1 | - | 9 | 3024 | |||
AK129843.1 | - | 2368 | ||||
AK130767.1 | - | 7 | 1798 | |||
AK291841.1 | - | 22 | 2484 | |||
AK308003.1 | - | 22 | 2475 | |||
AK309692.1 | - | 15 | 1812 | |||
AL521453.3 | - | 6 | 686 | |||
BC002536.1 | - | 22 | 2472 | 3140233 | ||
BC021698.1 | - | 13 | 1425 | |||
BC029138.1 | - | 22 | 2512 | 5180268 | ||
BC043156.1 | - | 6 | 2208 | |||
D25328.1 | - | 22 | 2469 | |||
M64784.1 | - | 8 | 986 |