Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321378.1 | NP_001308307.1 | 3 | 919 | ||
NM_002645.3 | NP_002636.2 | 2 | 850 | |||
XM_011520186.2 | XP_011518488.1 | 2 | 845 | |||
XM_017017924.1 | XP_016873413.1 | 3 | 866 | |||
GenBank mRNA | AK299474.1 | - | 2 | 782 | ||
AK304262.1 | - | 2 | 802 | |||
AK310248.1 | - | 2 | 840 | |||
BC113658.1 | - | 1 | 702 | 8322710 | ||
Y13367.1 | - | 1 | 636 |