Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321378.1 | NP_001308307.1 | 16 | 2871 | ||
NM_001321380.1 | NP_001308309.1 | 14 | 1672 | |||
NM_002645.3 | NP_002636.2 | 15 | 2802 | |||
XM_011520186.2 | XP_011518488.1 | 14 | 2629 | |||
XM_017017924.1 | XP_016873413.1 | 16 | 2818 | |||
GenBank mRNA | AK294416.1 | - | 14 | 1659 | ||
BC113658.1 | - | 14 | 2654 | 8322710 | ||
BX648778.1 | - | 5 | 738 | |||
Y13367.1 | - | 14 | 2588 |