Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 2, 3 | 1403 | ||
XM_011511687.1 | XP_011509989.1 | 2, 3 | 1420 | |||
GenBank mRNA | AK292430.1 | - | 2, 3 | 667 | ||
BC052985.2 | - | 2, 3 | 829 | 6149698 | ||
D50516.1 | - | 2, 3 | 663 | |||
U20165.1 | - | 2, 3 | 291 | |||
Z48923.1 | - | 2, 3 | 636 |