Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002646.3 | NP_002637.3 | 29 | 4717 | ||
XM_005245257.2 | XP_005245314.1 | 28 | 4782 | |||
XM_005245258.3 | XP_005245315.1 | 29 | 4701 | |||
XM_011509630.2 | XP_011507932.1 | 30 | 5576 | |||
XM_011509631.2 | XP_011507933.1 | 29 | 4893 | |||
XM_017001473.1 | XP_016856962.1 | 29 | 5712 | |||
XM_017001474.1 | XP_016856963.1 | 29 | 4915 | |||
XM_017001475.1 | XP_016856964.1 | 27 | 3663 | |||
GenBank mRNA | AB210006.1 | - | 27 | 4271 | ||
BC132876.1 | - | 29 | 4689 | 40146898 | ||
BC136645.1 | - | 29 | 4689 | 9020635 | ||
BC144342.1 | - | 30 | 4605 | 9052862 | ||
Y11312.1 | - | 29 | 4753 | |||
Y13892.1 | - | 27 | 4254 |