Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308020.1 | NP_001294949.1 | 13, 14 | 1492 | ||
NM_002647.3 | NP_002638.2 | 14, 15 | 1681 | |||
XM_011526031.2 | XP_011524333.1 | 15, 16 | 1846 | |||
GenBank mRNA | AK090567.1 | - | 1, 2 | 2215 | ||
AK298517.1 | - | 13, 14 | 1436 | |||
BC033004.2 | - | 14, 15 | 1631 | 4823337 | ||
BC053651.1 | - | 14, 15 | 1611 | 5517878 | ||
Z46973.1 | - | 14, 15 | 1634 |