Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206796.2 | NP_001193725.1 | 12 | 3048 | ||
NM_001206797.2 | NP_001193726.1 | 10 | 2489 | |||
NM_001206798.2 | NP_001193727.1 | 11 | 2736 | |||
NM_001206799.1 | NP_001193728.1 | 11 | 2378 | |||
NM_001316318.1 | NP_001303247.1 | 11 | 2931 | |||
NM_002654.5 | NP_002645.3 | 11 | 2711 | |||
NM_182470.3 | NP_872270.1 | 11 | 2927 | |||
NM_182471.3 | NP_872271.1 | 11 | 2711 | |||
XM_005254443.1 | XP_005254500.1 | 12 | 2435 | |||
XM_005254445.4 | XP_005254502.1 | 11 | 2272 | |||
XM_006720570.1 | XP_006720633.1 | 12 | 2532 | |||
XM_011521670.2 | XP_011519972.1 | 11 | 2497 | |||
XM_017022313.1 | XP_016877802.1 | 11 | 2272 | |||
GenBank mRNA | AF025439.1 | - | 4 | 1089 | ||
AK092369.1 | - | 11 | 2461 | |||
AK297951.1 | - | 2741 | ||||
AK299413.1 | - | 11 | 2215 | |||
BC000481.2 | - | 11 | 2293 | 2964687 | ||
BC007640.1 | - | 11 | 2277 | 3533876 | ||
BC007952.2 | - | 11 | 2289 | 4299213 | ||
BC012811.2 | - | 11 | 2289 | 2958817 | ||
BC019265.2 | - | 6 | 1625 | |||
BC023328.1 | - | 7 | 1661 | |||
BC023592.2 | - | 11 | 2239 | |||
BC035198.1 | - | 11 | 2243 | 3859987 | ||
BC094767.1 | - | 8 | 1838 | 5296639 | ||
BC096823.1 | - | 11 | 2478 | |||
BX648597.1 | - | 2 | 2281 | |||
EU794631.1 | - | 11 | 2430 | |||
M26252.1 | - | 11 | 2278 |