Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM981706.1 | - | 1 | 147 |
Gene Symbol: PLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002668.2 | NP_002659.1 | 5 | 861 | ||
GenBank mRNA | L09604.1 | - | 5 | 772 |