Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001998.2 | NP_001001998.1 | 22 | 2479 | ||
NM_002685.3 | NP_002676.1 | 21 | 2404 | |||
XM_005263475.2 | XP_005263532.2 | 22 | 2453 | |||
XM_017001494.1 | XP_016856983.1 | 21 | 2179 | |||
XM_017001495.1 | XP_016856984.1 | 21 | 2164 | |||
XM_017001496.1 | XP_016856985.1 | 22 | 2524 | |||
XM_017001497.1 | XP_016856986.1 | 22 | 2509 | |||
GenBank mRNA | AB209236.1 | - | 18 | 4011 | ||
AK093955.1 | - | 2 | 2211 | |||
AK123365.1 | - | 10 | 1652 | |||
AK296559.1 | - | 20 | 2979 | |||
BC009908.2 | - | 18 | 1801 | |||
BC039901.1 | - | 22 | 2420 | 5505500 | ||
BC073788.1 | - | 22 | 2455 | 5420763 | ||
L01457.1 | - | 21 | 2363 | |||
X66113.1 | - | 22 | 2437 |