Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144885.1 | NP_001138357.1 | 4 | 715 | ||
NM_001144886.1 | NP_001138358.1 | 3 | 667 | |||
NM_001144887.1 | NP_001138359.1 | 3 | 594 | |||
NM_004143.3 | NP_004134.2 | 3 | 1015 | |||
XM_011530958.1 | XP_011529260.1 | 4 | 706 | |||
GenBank mRNA | AB451286.1 | - | 2 | 439 | ||
AB451418.1 | - | 2 | 439 | |||
BC004240.1 | - | 3 | 968 | 3627230 | ||
U65092.1 | - | 3 | 634 |