Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004147.3 | NP_004138.1 | 8 | 1037 | ||
GenBank mRNA | AF078103.1 | - | 8 | 967 | ||
AJ005940.1 | - | 8 | 971 | |||
AK098576.1 | - | 8 | 913 | |||
AK315659.1 | - | 8 | 995 | |||
AL133085.1 | - | 3 | 256 | |||
BC019285.1 | - | 8 | 976 | 2823263 | ||
BC020803.1 | - | 8 | 971 | 4250998 | ||
BT007237.1 | - | 8 | 906 | |||
CR456488.1 | - | 8 | 948 | |||
CR542059.1 | - | 8 | 906 |