Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301020.1 | NP_001287949.1 | 3 | 495 | ||
NM_004152.3 | NP_004143.1 | 3 | 501 | |||
GenBank mRNA | AF090094.1 | - | 1428 | |||
AK307221.1 | - | 3 | 467 | |||
AL542477.3 | - | 1 | 73 | |||
BC010063.1 | - | 3 | 451 | |||
BI667301.1 | - | 3 | 472 | |||
BM423450.1 | - | 3 | 450 | |||
D78361.1 | - | 3 | 532 | |||
D87914.1 | - | 3 | 448 | |||
U09202.1 | - | 3 | 433 |