Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
ASPH
Gene Name:
aspartate beta-hydroxylase
Gene Aliases:
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location:
Chr.8: 61500556 - 61714640 on Build GRCh38
UniGene ID:
Hs.332422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14391
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASPH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164750.1
NP_001158222.1
2
200
NM_001164751.1
NP_001158223.1
2
200
NM_001164752.1
NP_001158224.1
2
200
NM_001164753.1
NP_001158225.1
2
200
NM_001164754.1
NP_001158226.1
2
395
NM_001164755.1
NP_001158227.1
2
395
NM_001164756.1
NP_001158228.1
2
395
NM_004318.3
NP_004309.2
2
395
NM_020164.4
NP_064549.1
2
200
NM_032466.3
NP_115855.1
2
395
NM_032467.3
NP_115856.1
2
200
NM_032468.4
NP_115857.1
2
200
XM_005251235.2
XP_005251292.1
2
364
XM_005251236.2
XP_005251293.1
2
364
XM_005251238.2
XP_005251295.1
2
364
XM_005251239.2
XP_005251296.1
2
364
XM_005251240.1
XP_005251297.1
2
142
XM_005251242.2
XP_005251299.1
2
364
XM_005251243.2
XP_005251300.1
2
364
XM_005251244.1
XP_005251301.1
2
142
XM_005251246.2
XP_005251303.1
2
364
XM_005251247.2
XP_005251304.1
2
364
XM_005251248.1
XP_005251305.1
2
50
XM_005251250.2
XP_005251307.1
2
364
XM_017013419.1
XP_016868908.1
2
364
XM_017013420.1
XP_016868909.1
2
364
XM_017013421.1
XP_016868910.1
2
364
XM_017013422.1
XP_016868911.1
2
364
XM_017013423.1
XP_016868912.1
2
364
XM_017013424.1
XP_016868913.1
2
364
XM_017013425.1
XP_016868914.1
2
364
XM_017013426.1
XP_016868915.1
2
142
XM_017013427.1
XP_016868916.1
2
142
XM_017013428.1
XP_016868917.1
2
364
XM_017013429.1
XP_016868918.1
2
364
XM_017013430.1
XP_016868919.1
2
364
XM_017013431.1
XP_016868920.1
2
50
XM_017013432.1
XP_016868921.1
2
364
XM_017013433.1
XP_016868922.1
2
50
XM_017013434.1
XP_016868923.1
2
364
XM_017013435.1
XP_016868924.1
2
142
XM_017013436.1
XP_016868925.1
2
364
XM_017013437.1
XP_016868926.1
2
364
XM_017013438.1
XP_016868927.1
2
364
XM_017013439.1
XP_016868928.1
2
50
XM_017013440.1
XP_016868929.1
2
142
XM_017013441.1
XP_016868930.1
2
364
XM_017013442.1
XP_016868931.1
2
364
XM_017013443.1
XP_016868932.1
2
364
XM_017013444.1
XP_016868933.1
2
50
XM_017013445.1
XP_016868934.1
2
50
XM_017013446.1
XP_016868935.1
2
50
XM_017013447.1
XP_016868936.1
2
50
GenBank mRNA
AF184241.1
-
2
80
AF224468.1
-
2
150
AF224469.1
-
2
125
AF289489.1
-
2
137
AF306765.1
-
2
121
AK291836.1
-
2
208
AK295528.1
-
2
164
AK304314.1
-
2
156
BC015518.2
-
2
314
BC025236.1
-
2
323
BC066929.1
-
2
291
BC142967.1
-
2
272
BC144362.1
-
2
130
BC144363.1
-
2
131
CR996516.1
-
2
395
DA136318.1
-
2
364
DN993915.1
-
2
208
EU718494.1
-
2
203
FJ461473.1
-
2
126
S83325.1
-
2
137
U03109.1
-
2
203
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
pattern specification process
negative regulation of cell proliferation
regulation of protein stability
ion transmembrane transport
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
oxidation-reduction process
palate development
face morphogenesis
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
regulation of cardiac conduction
peptide-aspartate beta-dioxygenase activity
structural molecule activity
calcium ion binding
protein binding
structural constituent of muscle
electron carrier activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline