Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004560.3 | NP_004551.2 | 9 | 3776 | ||
XM_005252008.4 | XP_005252065.1 | 9 | 3741 | |||
XM_005252009.3 | XP_005252066.1 | 3 | 2482 | |||
XM_006717121.3 | XP_006717184.1 | 9 | 4398 | |||
XM_017014762.1 | XP_016870251.1 | 9 | 5473 | |||
XM_017014763.1 | XP_016870252.1 | 9 | 3652 | |||
GenBank mRNA | AB209154.1 | - | 11 | 4041 | ||
M97639.1 | - | 9 | 3776 |