Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004561.3 | NP_004552.2 | 4 | 2221 | ||
XM_005274018.3 | XP_005274075.1 | 4 | 2039 | |||
XM_011545067.2 | XP_011543369.1 | 4 | 2037 | |||
XM_011545068.2 | XP_011543370.1 | 4 | 2582 | |||
XM_017017837.1 | XP_016873326.1 | 4 | 2875 | |||
XM_017017838.1 | XP_016873327.1 | 4 | 1914 | |||
GenBank mRNA | BC059408.1 | - | 4 | 2197 | 30343789 | |
BX648725.1 | - | 4 | 2101 |