Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302777.1 | NP_001289706.1 | 4 | 948 | ||
NM_004332.3 | NP_004323.2 | 3 | 720 | |||
NR_026648.2 | 4 | 948 | ||||
NR_026649.2 | 3 | 720 | ||||
GenBank mRNA | AJ617684.1 | - | 3 | 368 | ||
AK297518.1 | - | 3 | 404 | |||
AK314909.1 | - | 4 | 602 | |||
AL832502.1 | - | 3 | 371 | |||
BC037778.1 | - | 4 | 953 | 4799692 | ||
BC106901.1 | - | 3 | 435 | 40031474 | ||
DC352501.1 | - | 3 | 404 | |||
X81372.1 | - | 4 | 516 |