Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005012.3 | NP_005003.2 | 8 | 1729 | ||
XM_011541526.1 | XP_011539828.1 | 8 | 1328 | |||
XM_017001376.1 | XP_016856865.1 | 7 | 1629 | |||
XM_017001377.1 | XP_016856866.1 | 10 | 4272 | |||
GenBank mRNA | AK308088.1 | - | 8 | 1735 | ||
AK309189.1 | - | 5 | 971 | |||
BC128386.1 | - | 9 | 1339 | |||
M97675.1 | - | 8 | 1705 |