Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004335.3 | NP_004326.1 | 1 | 326 | ||
GenBank mRNA | AK223124.1 | - | 1 | 264 | ||
AK291099.1 | - | 1 | 271 | |||
AK303593.1 | - | 1 | 296 | |||
BC033873.1 | - | 1 | 326 | 5217945 | ||
D28137.1 | - | 1 | 254 |