Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098620.2 | NP_001092090.1 | 13 | 1284 | ||
NM_001128911.1 | NP_001122383.1 | 14 | 1386 | |||
NM_001128912.1 | NP_001122384.1 | 14 | 1374 | |||
NM_001128913.1 | NP_001122385.1 | 13 | 1296 | |||
NM_001128914.1 | NP_001122386.1 | 12 | 1245 | |||
NM_005016.5 | NP_005007.2 | 14 | 1389 | |||
NM_031989.4 | NP_114366.1 | 14 | 1377 | |||
GenBank mRNA | AB188306.1 | - | 13 | 1284 | ||
AB208825.1 | - | 13 | 1108 | |||
AK023529.1 | - | 9 | 1988 | |||
AK123458.1 | - | 2 | 1290 | |||
AK130583.1 | - | 6 | 492 | |||
AK225357.1 | - | 11 | 959 | |||
AK292141.1 | - | 14 | 1154 | |||
AK299210.1 | - | 10 | 845 | |||
AK302067.1 | - | 11 | 974 | |||
BC001155.1 | - | 13 | 1115 | 3356341 | ||
BC035420.1 | - | 14 | 1362 | |||
BC071942.1 | - | 14 | 1379 | 6424433 | ||
BC107688.1 | - | 12 | 962 | 5395926 | ||
BE748080.1 | - | 122 | ||||
X78136.1 | - | 13 | 1058 |