Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191056.3 | NP_001177985.1 | 5 | 949 | ||
NM_001191057.3 | NP_001177986.1 | 5 | 949 | |||
NM_001191058.3 | NP_001177987.2 | 6 | 1329 | |||
NM_001191059.3 | NP_001177988.1 | 6 | 940 | |||
NM_001322055.1 | NP_001308984.1 | 6 | 1175 | |||
NM_001322056.1 | NP_001308985.1 | 6 | 940 | |||
NM_001322057.1 | NP_001308986.1 | 6 | 1175 | |||
NM_001322058.1 | NP_001308987.1 | 6 | 1329 | |||
NM_001322059.1 | NP_001308988.1 | 6 | 978 | |||
NM_005020.4 | NP_005011.1 | 6 | 713 | |||
XM_017012264.1 | XP_016867753.1 | 6 | 780 | |||
XM_017012265.1 | XP_016867754.1 | 6 | 631 | |||
XM_017012266.1 | XP_016867755.1 | 6 | 780 | |||
XM_017012267.1 | XP_016867756.1 | 6 | 685 | |||
GenBank mRNA | AK056170.1 | - | 5 | 936 | ||
AK091734.1 | - | 6 | 1254 | |||
AK094180.1 | - | 6 | 872 | |||
BC022479.1 | - | 5 | 950 | 4795813 | ||
BC022525.1 | - | 5 | 950 | 4795158 | ||
U40371.1 | - | 6 | 656 | |||
U40372.1 | - | 1 | 46 |